siRNA Details
Gene Symbol: PLEKHH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020715.2 | NP_065766.1 | 23 | 3416 | ||
XM_011537009.1 | XP_011535311.1 | 23 | 3592 | |||
XM_011537015.2 | XP_011535317.1 | 16 | 3647 | |||
XM_017021499.1 | XP_016876988.1 | 22 | 3573 | |||
XM_017021500.1 | XP_016876989.1 | 23 | 3508 | |||
XM_017021501.1 | XP_016876990.1 | 22 | 3489 | |||
XM_017021502.1 | XP_016876991.1 | 16 | 3369 | |||
XM_017021503.1 | XP_016876992.1 | 16 | 2729 | |||
XM_017021504.1 | XP_016876993.1 | 18 | 2871 | |||
GenBank mRNA | AB033026.1 | - | 23 | 3392 | ||
AK295896.1 | - | 16 | 2116 | |||
BC146787.1 | - | 23 | 3392 | 9007246 | ||
BX648137.1 | - | 1 | 896 | |||
BX648464.1 | - | 14 | 2528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
