siRNA Details
Gene Symbol: PLEKHH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020715.2 | NP_065766.1 | 29 | 4188 | ||
XM_011537009.1 | XP_011535311.1 | 29 | 4364 | |||
XM_011537015.2 | XP_011535317.1 | 22 | 4419 | |||
XM_017021499.1 | XP_016876988.1 | 28 | 4345 | |||
XM_017021500.1 | XP_016876989.1 | 29 | 4280 | |||
XM_017021501.1 | XP_016876990.1 | 28 | 4261 | |||
XM_017021502.1 | XP_016876991.1 | 22 | 4141 | |||
XM_017021503.1 | XP_016876992.1 | 22 | 3501 | |||
XM_017021504.1 | XP_016876993.1 | 24 | 3643 | |||
GenBank mRNA | AB033026.1 | - | 29 | 4164 | ||
AK123262.1 | - | 2 | 652 | |||
AK124787.1 | - | 6 | 1245 | |||
AK293950.1 | - | 12 | 1458 | |||
AK295896.1 | - | 22 | 2888 | |||
BC014159.2 | - | 6 | 647 | |||
BC146787.1 | - | 29 | 4164 | 9007246 | ||
BX648137.1 | - | 6 | 3520 | |||
BX648464.1 | - | 20 | 3300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
