siRNA Details
Gene Symbol: ZNF599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007248.2 | NP_001007249.1 | 4 | 971 | ||
XM_017026369.1 | XP_016881858.1 | 4 | 1046 | |||
GenBank mRNA | AK055225.1 | - | 4 | 957 | ||
BC044615.1 | - | 5 | 1087 | 5266705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
