siRNA Details
Gene Symbol: YBEY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006114.2 | NP_001006114.1 | 2 | 290 | ||
NM_001314022.1 | NP_001300951.1 | 2 | 612 | |||
NM_001314025.1 | NP_001300954.1 | 2 | 290 | |||
NM_001314026.1 | NP_001300955.1 | 2 | 612 | |||
NM_058181.2 | NP_478061.1 | 2 | 612 | |||
XM_005261156.3 | XP_005261213.2 | 2 | 564 | |||
XM_011529629.2 | XP_011527931.1 | 2 | 260 | |||
XM_011529630.2 | XP_011527932.1 | 2 | 291 | |||
XM_011529631.2 | XP_011527933.1 | 2 | 260 | |||
XM_011529633.2 | XP_011527935.1 | 2 | 260 | |||
XM_011529634.2 | XP_011527936.2 | 2 | 564 | |||
XM_011529635.2 | XP_011527937.1 | 2 | 291 | |||
XM_017028394.1 | XP_016883883.1 | 2 | 564 | |||
XM_017028396.1 | XP_016883885.1 | 307 | ||||
GenBank mRNA | AK128875.1 | - | 2 | 224 | ||
AK294975.1 | - | 2 | 271 | |||
AY040873.1 | - | 2 | 558 | |||
AY040874.1 | - | 2 | 558 | |||
BC022828.1 | - | 2 | 231 | 3928883 | ||
BC119770.1 | - | 2 | 236 | 40108760 | ||
BC122865.1 | - | 2 | 236 | 40108759 | ||
BC131553.1 | - | 2 | 236 | 40089560 | ||
BG473705.1 | - | 1 | 196 | |||
BM796288.1 | - | 2 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
