siRNA Details
Gene Symbol: OR52E8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005168.1 | NP_001005168.1 | 1 | 349 | ||
GenBank mRNA | BC137030.1 | - | 1 | 381 | 9021026 | |
BC137041.1 | - | 1 | 381 | 9021037 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
