siRNA Details
Gene Symbol: NPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099456.2 | NP_001092926.2 | 1 | 516 | ||
GenBank mRNA | AB084276.1 | - | 1 | 296 | ||
AL365521.1 | - | 1 | 491 | |||
BC150534.1 | - | 1 | 20 | |||
BC171886.1 | - | 1 | 733 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
