siRNA Details
Gene Symbol: IGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000612.5 | NP_000603.1 | 4 | 4634 | ||
NM_001007139.5 | NP_001007140.2 | 5 | 4608 | |||
NM_001127598.2 | NP_001121070.1 | 5 | 4321 | |||
NM_001291861.2 | NP_001278790.1 | 4 | 3985 | |||
NM_001291862.2 | NP_001278791.1 | 4 | 4156 | |||
GenBank mRNA | AF318382.1 | - | 1 | 2085 | ||
AK025719.1 | - | 1 | 1754 | |||
AY971350.1 | - | 1 | 135 | |||
BC011786.2 | - | 1 | 152 | |||
BC042127.1 | - | 2 | 377 |
Gene Symbol: INS-IGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003512.3 | 7 | 4596 | |||
GenBank mRNA | BF205917.1 | - | 1 | 581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
