siRNA Details
Gene Symbol: CAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001234.4 | NP_001225.1 | 2 | 291 | ||
NM_033337.2 | NP_203123.1 | 2 | 291 | |||
GenBank mRNA | AF036365.1 | - | 2 | 261 | ||
AF043101.1 | - | 2 | 211 | |||
AK291892.1 | - | 2 | 291 | |||
BC069368.1 | - | 2 | 264 | 7262168 | ||
BC102033.1 | - | 2 | 251 | 40033505 | ||
BC102036.1 | - | 2 | 251 | 40033114 | ||
BC102037.1 | - | 2 | 251 | 40033118 | ||
Y14747.1 | - | 2 | 225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
