siRNA Details
Gene Symbol: FGF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321931.1 | NP_001308860.1 | 6 | 997 | ||
NM_001321932.1 | NP_001308861.1 | 6 | 1083 | |||
NM_001321933.1 | NP_001308862.1 | 6 | 993 | |||
NM_001321934.1 | NP_001308863.1 | 5 | 756 | |||
NM_001321935.1 | NP_001308864.1 | 5 | 609 | |||
NM_001321936.1 | NP_001308865.1 | 6 | 936 | |||
NM_001321938.1 | NP_001308867.1 | 7 | 984 | |||
NM_001321939.1 | NP_001308868.1 | 4 | 991 | |||
NM_001321940.1 | NP_001308869.1 | 6 | 1339 | |||
NM_001321941.1 | NP_001308870.1 | 6 | 1171 | |||
NM_001321942.1 | NP_001308871.1 | 6 | 1178 | |||
NM_001321943.1 | NP_001308872.1 | 6 | 1215 | |||
NM_001321944.1 | NP_001308873.1 | 6 | 1431 | |||
NM_001321945.1 | NP_001308874.1 | 6 | 974 | |||
NM_001321946.1 | NP_001308875.1 | 7 | 1080 | |||
NM_001321947.1 | NP_001308876.1 | 6 | 898 | |||
NM_001321948.1 | NP_001308877.1 | 6 | 970 | |||
NM_001321949.1 | NP_001308878.1 | 6 | 1667 | |||
NM_004115.3 | NP_004106.1 | 5 | 733 | |||
NM_175929.2 | NP_787125.1 | 5 | 844 | |||
GenBank mRNA | AY188178.1 | - | 5 | 844 | ||
BC100920.1 | - | 5 | 843 | 40003901 | ||
BC100921.1 | - | 5 | 843 | 40003902 | ||
BC100922.2 | - | 5 | 843 | 40003903 | ||
U66200.1 | - | 5 | 733 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
