siRNA Details
Gene Symbol: IFIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270927.1 | NP_001257856.1 | 3 | 896 | ||
NM_001270928.1 | NP_001257857.1 | 3 | 913 | |||
NM_001270929.1 | NP_001257858.1 | 3 | 908 | |||
NM_001270930.1 | NP_001257859.1 | 3 | 1034 | |||
NM_001548.4 | NP_001539.3 | 2 | 799 | |||
GenBank mRNA | AK092813.1 | - | 2 | 1827 | ||
AK308688.1 | - | 3 | 800 | |||
AK314588.1 | - | 2 | 722 | |||
AL548182.3 | - | 2 | 702 | |||
BC007091.1 | - | 2 | 724 | 4250452 | ||
BT006667.1 | - | 1 | 632 | |||
CD106320.1 | - | 2 | 675 | |||
X03557.1 | - | 2 | 696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
