siRNA Details
Gene Symbol: FAM87A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103537.1 | 3 | 2863 | |||
GenBank mRNA | BC037297.2 | - | 3 | 2862 | ||
BC118644.1 | - | 1 | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
