siRNA Details
Gene Symbol: EPHA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099439.1 | NP_001092909.1 | 9, 10 | 1912 | ||
XM_017001080.1 | XP_016856569.1 | 5, 6 | 654 | |||
XM_017001081.1 | XP_016856570.1 | 5, 6 | 654 | |||
GenBank mRNA | AK131215.1 | - | 5, 6 | 626 | ||
AK309089.1 | - | 5, 6 | 564 | |||
AY726560.1 | - | 3, 4 | 247 | |||
BC041360.1 | - | 5, 6 | 500 | |||
BC112933.1 | - | 3, 4 | 326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
