siRNA Details
Gene Symbol: IRF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001572.3 | NP_001563.2 | 11 | 1853 | ||
NM_004029.2 | NP_004020.1 | 10 | 1766 | |||
NM_004031.2 | NP_004022.2 | 9 | 1932 | |||
XM_005252906.3 | XP_005252963.1 | 10 | 1918 | |||
XM_005252907.3 | XP_005252964.1 | 10 | 1915 | |||
XM_005252909.3 | XP_005252966.1 | 9 | 1831 | |||
XM_011520066.2 | XP_011518368.1 | 11 | 1829 | |||
XM_017017674.1 | XP_016873163.1 | 4 | 731 | |||
GenBank mRNA | AF076494.1 | - | 9 | 1857 | ||
AK303752.1 | - | 9 | 1664 | |||
BC136555.1 | - | 9 | 1568 | 9020545 | ||
KC477210.1 | - | 8 | 1845 | |||
U53830.1 | - | 10 | 1775 | |||
U53831.1 | - | 9 | 1688 | |||
U53832.1 | - | 9 | 1607 | |||
U73036.1 | - | 11 | 1792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
