siRNA Details
Gene Symbol: IRF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001572.3 | NP_001563.2 | 9, 10 | 1601 | ||
NM_004029.2 | NP_004020.1 | 8, 9 | 1514 | |||
NM_004031.2 | NP_004022.2 | 7, 8 | 1680 | |||
XM_005252906.3 | XP_005252963.1 | 8, 9 | 1666 | |||
XM_005252907.3 | XP_005252964.1 | 8, 9 | 1663 | |||
XM_005252909.3 | XP_005252966.1 | 7, 8 | 1579 | |||
XM_011520066.2 | XP_011518368.1 | 9, 10 | 1577 | |||
XM_017017674.1 | XP_016873163.1 | 2, 3 | 479 | |||
GenBank mRNA | AK303752.1 | - | 7, 8 | 1412 | ||
BC136555.1 | - | 7, 8 | 1316 | 9020545 | ||
KC477210.1 | - | 6, 7 | 1593 | |||
U53830.1 | - | 8, 9 | 1523 | |||
U53831.1 | - | 7, 8 | 1436 | |||
U53832.1 | - | 7, 8 | 1355 | |||
U73036.1 | - | 9, 10 | 1540 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
