siRNA Details
Gene Symbol: F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000132.3 | NP_000123.1 | 25 | 6978 | ||
NM_019863.2 | NP_063916.1 | 4 | 547 | |||
GenBank mRNA | AK289947.1 | - | 4 | 477 | ||
AK313707.1 | - | 4 | 432 | |||
AX746360.1 | - | 25 | 6807 | |||
BC022513.1 | - | 4 | 466 | |||
BC064380.1 | - | 4 | 526 | |||
BC098389.1 | - | 4 | 545 | |||
BC111967.1 | - | 4 | 435 | |||
BC111969.1 | - | 4 | 434 | |||
BI547023.1 | - | 4 | 588 | |||
K01740.1 | - | 25 | 6957 | |||
KT878539.1 | - | 2 | 228 | |||
M14113.1 | - | 25 | 6978 | |||
M90707.1 | - | 4 | 442 | |||
X01179.1 | - | 25 | 6916 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
