siRNA Details
Gene Symbol: F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000132.3 | NP_000123.1 | 26 | 7144 | ||
NM_019863.2 | NP_063916.1 | 5 | 713 | |||
GenBank mRNA | AK289947.1 | - | 5 | 643 | ||
AK313707.1 | - | 5 | 598 | |||
AX746360.1 | - | 26 | 6973 | |||
BC022513.1 | - | 5 | 632 | |||
BC064380.1 | - | 5 | 692 | |||
BC098389.1 | - | 5 | 711 | |||
BC111967.1 | - | 5 | 601 | |||
BC111969.1 | - | 5 | 600 | |||
K01740.1 | - | 26 | 7123 | |||
KT878539.1 | - | 3 | 394 | |||
M14113.1 | - | 26 | 7144 | |||
M90707.1 | - | 5 | 608 | |||
X01179.1 | - | 26 | 7082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
