siRNA Details
Gene Symbol: F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000132.3 | NP_000123.1 | 26 | 8178 | ||
NM_019863.2 | NP_063916.1 | 5 | 1747 | |||
GenBank mRNA | BC022513.1 | - | 5 | 1666 | ||
BC064380.1 | - | 5 | 1726 | |||
BC098389.1 | - | 5 | 1745 | |||
K01740.1 | - | 26 | 8155 | |||
M14113.1 | - | 26 | 8178 | |||
M90707.1 | - | 5 | 1642 | |||
X01179.1 | - | 26 | 8116 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
