siRNA Details
Gene Symbol: ZNF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016265.3 | NP_057349.2 | 5 | 985 | ||
GenBank mRNA | AB021643.1 | - | 5 | 985 | ||
AF505656.1 | - | 5 | 652 | |||
AK293925.1 | - | 6 | 631 | |||
AY500354.1 | - | 2 | 287 | |||
BC137068.1 | - | 4 | 477 | 9021065 | ||
BC137069.1 | - | 4 | 477 | 9021066 | ||
BC144465.1 | - | 5 | 530 | |||
BC154414.1 | - | 5 | 973 | 40148925 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
