siRNA Details
Gene Symbol: CHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005271.2 | NP_001005271.2 | 12 | 2209 | ||
NM_001005273.2 | NP_001005273.1 | 12 | 2180 | |||
NM_005852.3 | NP_005843.2 | 12 | 2180 | |||
XM_005256427.4 | XP_005256484.1 | 12 | 2250 | |||
XM_005256428.4 | XP_005256485.1 | 12 | 2250 | |||
XM_005256429.4 | XP_005256486.1 | 12 | 2250 | |||
XM_005256431.4 | XP_005256488.1 | 12 | 2250 | |||
XM_006721423.3 | XP_006721486.1 | 12 | 2250 | |||
XM_006721424.3 | XP_006721487.1 | 12 | 2225 | |||
XM_006721428.3 | XP_006721491.1 | 12 | 2250 | |||
XM_017024061.1 | XP_016879550.1 | 12 | 2232 | |||
XM_017024062.1 | XP_016879551.1 | 12 | 2207 | |||
XM_017024063.1 | XP_016879552.1 | 12 | 2165 | |||
XM_017024064.1 | XP_016879553.1 | 12 | 2225 | |||
XM_017024065.1 | XP_016879554.1 | 11 | 2046 | |||
XM_017024066.1 | XP_016879555.1 | 13 | 1985 | |||
XM_017024067.1 | XP_016879556.1 | 13 | 2168 | |||
XM_017024068.1 | XP_016879557.1 | 12 | 2141 | |||
XM_017024069.1 | XP_016879558.1 | 12 | 2032 | |||
XM_017024070.1 | XP_016879559.1 | 12 | 2207 | |||
GenBank mRNA | U91543.1 | - | 12 | 2180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
