siRNA Details
Gene Symbol: CD86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206924.1 | NP_001193853.1 | 6 | 1165 | ||
NM_001206925.1 | NP_001193854.1 | 6 | 1451 | |||
NM_006889.4 | NP_008820.3 | 7 | 1556 | |||
NM_175862.4 | NP_787058.4 | 7 | 1501 | |||
NM_176892.1 | NP_795711.1 | 6 | 1412 | |||
GenBank mRNA | AK291497.1 | - | 7 | 1409 | ||
AK294663.1 | - | 6 | 1160 | |||
AK301237.1 | - | 6 | 1446 | |||
AK316203.1 | - | 6 | 1446 | |||
BC040261.1 | - | 7 | 1488 | 5173789 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
