siRNA Details
Gene Symbol: CTAGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172241.2 | NP_758441.2 | 1 | 3137 | ||
GenBank mRNA | AF177229.1 | - | 1 | 89 | ||
AF273058.1 | - | 1 | 3144 | |||
BC132871.1 | - | 1 | 3070 | 40146893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
