siRNA Details
Gene Symbol: HCAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006018.2 | NP_006009.2 | 1 | 480 | ||
GenBank mRNA | AK290930.1 | - | 1 | 361 | ||
AK313212.1 | - | 1 | 379 | |||
BC038955.1 | - | 1 | 370 | |||
BC047891.1 | - | 1 | 385 | 5750043 | ||
BI908162.1 | - | 1 | 373 | |||
D10923.1 | - | 1 | 385 | |||
DA953971.1 | - | 1 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
