siRNA Details
Gene Symbol: SOCS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014598.3 | NP_055413.1 | 8 | 1650 | ||
XM_017024551.1 | XP_016880040.1 | 7 | 1796 | |||
XM_017024552.1 | XP_016880041.1 | 6 | 1691 | |||
GenBank mRNA | AA233668.1 | - | 2 | 90 | ||
AB005216.1 | - | 8 | 1161 | |||
BC121031.1 | - | 8 | 1080 | |||
BC128607.1 | - | 8 | 1106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
