siRNA Details
Gene Symbol: SOCS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014598.3 | NP_055413.1 | 6 | 1367 | ||
XM_017024551.1 | XP_016880040.1 | 5 | 1513 | |||
XM_017024552.1 | XP_016880041.1 | 4 | 1408 | |||
GenBank mRNA | AB005216.1 | - | 6 | 878 | ||
BC121030.1 | - | 7 | 1574 | |||
BC121031.1 | - | 6 | 797 | |||
BC128607.1 | - | 6 | 823 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
