siRNA Details
Gene Symbol: ADGRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008701.2 | NP_001008701.1 | 24 | 6551 | ||
NM_014921.4 | NP_055736.2 | 23 | 6536 | |||
XM_005259818.3 | XP_005259875.1 | 23 | 6664 | |||
XM_011527796.2 | XP_011526098.1 | 24 | 6700 | |||
XM_011527798.2 | XP_011526100.1 | 23 | 6685 | |||
XM_011527801.2 | XP_011526103.1 | 22 | 6140 | |||
XM_017026475.1 | XP_016881964.1 | 24 | 6703 | |||
XM_017026476.1 | XP_016881965.1 | 25 | 6813 | |||
XM_017026477.1 | XP_016881966.1 | 24 | 6682 | |||
XM_017026478.1 | XP_016881967.1 | 24 | 6631 | |||
XM_017026479.1 | XP_016881968.1 | 23 | 6613 | |||
XM_017026480.1 | XP_016881969.1 | 21 | 6118 | |||
GenBank mRNA | AK022001.1 | - | 1 | 930 | ||
BC013275.2 | - | 1 | 888 | |||
BC019928.1 | - | 1 | 486 | |||
BC072445.1 | - | 1 | 893 | |||
BC090949.1 | - | 1 | 1044 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
