siRNA Details
Gene Symbol: MORF4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265603.1 | NP_001252532.1 | 10 | 1105 | ||
NM_001265604.1 | NP_001252533.1 | 12 | 1173 | |||
NM_001265605.1 | NP_001252534.1 | 12 | 1073 | |||
NM_006791.3 | NP_006782.1 | 12 | 1220 | |||
NM_206839.2 | NP_996670.1 | 13 | 1337 | |||
GenBank mRNA | AF070664.1 | - | 12 | 1027 | ||
AF100615.1 | - | 12 | 1114 | |||
AF109188.1 | - | 12 | 1009 | |||
AF131847.1 | - | 8 | 743 | |||
AF161546.1 | - | 12 | 1006 | |||
AF167173.1 | - | 12 | 1022 | |||
AK095823.1 | - | 13 | 1622 | |||
AK098485.1 | - | 12 | 1140 | |||
AK296650.1 | - | 13 | 1228 | |||
AK300789.1 | - | 12 | 1157 | |||
AL137697.1 | - | 12 | 1013 | |||
AY148481.1 | - | 13 | 1231 | |||
BC002936.2 | - | 12 | 1101 | 3955120 | ||
BC022845.2 | - | 12 | 1101 | 2988803 | ||
BC141840.1 | - | 12 | 1028 | 8860303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
