siRNA Details
Gene Symbol: MORF4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265603.1 | NP_001252532.1 | 8 | 822 | ||
NM_001265604.1 | NP_001252533.1 | 10 | 890 | |||
NM_001265605.1 | NP_001252534.1 | 10 | 790 | |||
NM_006791.3 | NP_006782.1 | 10 | 937 | |||
NM_206839.2 | NP_996670.1 | 11 | 1054 | |||
XM_017021872.1 | XP_016877361.1 | 10 | 902 | |||
GenBank mRNA | AF070664.1 | - | 10 | 744 | ||
AF100615.1 | - | 10 | 831 | |||
AF109188.1 | - | 10 | 726 | |||
AF131847.1 | - | 6 | 460 | |||
AF161546.1 | - | 10 | 723 | |||
AF167173.1 | - | 10 | 739 | |||
AF218011.1 | - | 10 | 842 | |||
AK095823.1 | - | 11 | 1339 | |||
AK098485.1 | - | 10 | 857 | |||
AK296650.1 | - | 11 | 945 | |||
AK300789.1 | - | 10 | 874 | |||
AL137697.1 | - | 10 | 730 | |||
AY148481.1 | - | 11 | 948 | |||
BC002936.2 | - | 10 | 818 | 3955120 | ||
BC022845.2 | - | 10 | 818 | 2988803 | ||
BC067826.1 | - | 10 | 713 | 4824022 | ||
BC121092.1 | - | 6 | 440 | 40119977 | ||
BC121093.1 | - | 6 | 440 | 40119978 | ||
BC141840.1 | - | 10 | 745 | 8860303 | ||
BX382045.2 | - | 10 | 734 | |||
CX165647.1 | - | 8 | 573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
