siRNA Details
Gene Symbol: HIST2H2AC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003517.2 | NP_003508.1 | 1 | 363 | ||
GenBank mRNA | BC060324.1 | - | 1 | 398 | 3892650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
