siRNA Details
Gene Symbol: XKR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318251.1 | NP_001305180.1 | 3 | 605 | ||
NM_175878.4 | NP_787074.2 | 3 | 602 | |||
XM_017028600.1 | XP_016884089.1 | 3 | 631 | |||
GenBank mRNA | AK098608.1 | - | 3 | 591 | ||
AY534240.1 | - | 3 | 595 | |||
AY989815.3 | - | 3 | 580 | |||
BC137521.1 | - | 3 | 569 | 9021521 | ||
BC137522.1 | - | 3 | 569 | 9021522 | ||
BC144679.1 | - | 3 | 572 | 9053213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
