siRNA Details
Gene Symbol: IL36RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012275.2 | NP_036407.1 | 5 | 1292 | ||
NM_173170.1 | NP_775262.1 | 5 | 1264 | |||
GenBank mRNA | AF186094.1 | - | 5 | 1293 | ||
AF201830.1 | - | 4 | 1158 | |||
AJ242737.1 | - | 5 | 1212 | |||
AJ242738.1 | - | 5 | 1203 | |||
AK002125.1 | - | 1 | 467 | |||
AY359117.1 | - | 5 | 1197 | |||
BC024747.1 | - | 5 | 1264 | 4996939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
