siRNA Details
Gene Symbol: IL36RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012275.2 | NP_036407.1 | 5 | 1113 | ||
NM_173170.1 | NP_775262.1 | 5 | 1085 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
