siRNA Details
Gene Symbol: SLC6A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010898.2 | NP_001010898.1 | 5 | 1048 | ||
XM_006710643.2 | XP_006710706.1 | 5 | 756 | |||
GenBank mRNA | AK289982.1 | - | 5 | 1044 | ||
BC140908.1 | - | 4 | 633 | 9056720 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
