siRNA Details
Gene Symbol: L3MBTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015478.6 | NP_056293.4 | 19 | 2990 | ||
NM_032107.4 | NP_115479.4 | 22 | 3161 | |||
GenBank mRNA | AK123176.1 | - | 3 | 2143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
