siRNA Details
Gene Symbol: L3MBTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015478.6 | NP_056293.4 | 19 | 2386 | ||
NM_032107.4 | NP_115479.4 | 22 | 2557 | |||
GenBank mRNA | AK123176.1 | - | 3 | 1539 | ||
AK299199.1 | - | 22 | 2527 | |||
AL110279.1 | - | 10 | 3196 | |||
BC039820.1 | - | 19 | 2301 | 5199689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
