siRNA Details
Gene Symbol: L3MBTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015478.6 | NP_056293.4 | 2 | 397 | ||
NM_032107.4 | NP_115479.4 | 5 | 568 | |||
GenBank mRNA | AK299199.1 | - | 5 | 538 | ||
AK308334.1 | - | 2 | 364 | |||
AK310345.1 | - | 2 | 397 | |||
BC039820.1 | - | 2 | 312 | 5199689 | ||
DA204976.1 | - | 4 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
