siRNA Details
Gene Symbol: TTTY14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001543.3 | 1 | 197 | |||
NR_125733.1 | 1 | 197 | ||||
NR_125734.1 | 1 | 197 | ||||
NR_125735.1 | 1 | 197 | ||||
GenBank mRNA | DA411985.1 | - | 1 | 197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
