siRNA Details
Gene Symbol: TTTY14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001543.3 | 1 | 247 | |||
NR_125733.1 | 1 | 247 | ||||
NR_125734.1 | 1 | 247 | ||||
NR_125735.1 | 1 | 247 | ||||
GenBank mRNA | BI824313.1 | - | 1 | 41 | ||
DA411985.1 | - | 1 | 247 | |||
DB155066.1 | - | 1 | 240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
