siRNA Details
Gene Symbol: SLC35D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008783.2 | NP_001008783.1 | 2 | 1424 | ||
GenBank mRNA | AK295424.1 | - | 2 | 1273 | ||
AK316095.1 | - | 2 | 1424 | |||
BC067217.1 | - | 1 | 657 | |||
BC087842.1 | - | 2 | 1401 | 6191889 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
