siRNA Details
Gene Symbol: TCHHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008536.1 | NP_001008536.1 | 3 | 1316 | ||
GenBank mRNA | AY456639.1 | - | 3 | 1316 | ||
BC137493.1 | - | 3 | 1306 | 9021493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
