siRNA Details
Gene Symbol: ACTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199954.1 | NP_001186883.1 | 6 | 1490 | ||
NM_001614.3 | NP_001605.1 | 6 | 1371 | |||
NR_037688.1 | 6 | 1371 | ||||
GenBank mRNA | AK291937.1 | - | 6 | 1304 | ||
AK301175.1 | - | 7 | 1178 | |||
AK304637.1 | - | 7 | 1244 | |||
BC000292.2 | - | 6 | 1297 | 2819345 | ||
BC001920.1 | - | 6 | 1260 | 2820489 | ||
BC004223.2 | - | 5 | 1209 | |||
BC007442.2 | - | 6 | 1291 | 2962953 | ||
BC009544.1 | - | 3 | 585 | |||
BC009848.2 | - | 6 | 1288 | 3939552 | ||
BC010417.2 | - | 3 | 604 | |||
BC010999.2 | - | 6 | 1290 | 3940861 | ||
BC012050.1 | - | 6 | 1291 | 4554944 | ||
BC015005.1 | - | 6 | 1286 | 4855463 | ||
BC015695.2 | - | 6 | 1293 | 4842665 | ||
BC015779.1 | - | 6 | 1267 | 4856294 | ||
BC017450.1 | - | 5 | 1198 | |||
BC018774.1 | - | 6 | 1291 | 4869628 | ||
BC018861.2 | - | 3 | 731 | |||
BC023548.2 | - | 4 | 898 | |||
BC053572.1 | - | 6 | 1284 | 3865767 | ||
BC063495.1 | - | 5 | 1223 | |||
EU668331.1 | - | 3 | 803 | |||
M16247.1 | - | 3 | 803 | |||
X04098.1 | - | 6 | 1305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
