siRNA Details
Gene Symbol: ACTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199954.1 | NP_001186883.1 | 6 | 1418 | ||
NM_001614.3 | NP_001605.1 | 6 | 1299 | |||
NR_037688.1 | 6 | 1299 | ||||
GenBank mRNA | AK291937.1 | - | 6 | 1232 | ||
AK301175.1 | - | 7 | 1106 | |||
AK304637.1 | - | 7 | 1172 | |||
BC000292.2 | - | 6 | 1225 | 2819345 | ||
BC001920.1 | - | 6 | 1188 | 2820489 | ||
BC004223.2 | - | 5 | 1137 | |||
BC007442.2 | - | 6 | 1219 | 2962953 | ||
BC009544.1 | - | 3 | 513 | |||
BC009848.2 | - | 6 | 1216 | 3939552 | ||
BC010417.2 | - | 3 | 532 | |||
BC010999.2 | - | 6 | 1218 | 3940861 | ||
BC012050.1 | - | 6 | 1219 | 4554944 | ||
BC015005.1 | - | 6 | 1214 | 4855463 | ||
BC015695.2 | - | 6 | 1221 | 4842665 | ||
BC015779.1 | - | 6 | 1195 | 4856294 | ||
BC017450.1 | - | 5 | 1126 | |||
BC018774.1 | - | 6 | 1219 | 4869628 | ||
BC018861.2 | - | 3 | 659 | |||
BC023548.2 | - | 4 | 826 | |||
BC053572.1 | - | 6 | 1212 | 3865767 | ||
BC063495.1 | - | 5 | 1151 | |||
EU668331.1 | - | 3 | 731 | |||
M16247.1 | - | 3 | 731 | |||
X04098.1 | - | 6 | 1233 |
Gene Symbol: ACTG1P20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033926.1 | 1 | 1957 | |||
GenBank mRNA | AK056682.1 | - | 1 | 1957 | ||
BX648289.1 | - | 1 | 3435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
