siRNA Details
Gene Symbol: ACTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199954.1 | NP_001186883.1 | 6 | 1571 | ||
NM_001614.3 | NP_001605.1 | 6 | 1452 | |||
NR_037688.1 | 6 | 1452 | ||||
GenBank mRNA | AK291937.1 | - | 6 | 1385 | ||
AK301175.1 | - | 7 | 1259 | |||
AK304637.1 | - | 7 | 1325 | |||
BC000292.2 | - | 6 | 1378 | 2819345 | ||
BC001920.1 | - | 6 | 1341 | 2820489 | ||
BC004223.2 | - | 5 | 1290 | |||
BC007442.2 | - | 6 | 1372 | 2962953 | ||
BC009544.1 | - | 3 | 666 | |||
BC009848.2 | - | 6 | 1369 | 3939552 | ||
BC010999.2 | - | 6 | 1371 | 3940861 | ||
BC012050.1 | - | 6 | 1372 | 4554944 | ||
BC015005.1 | - | 6 | 1367 | 4855463 | ||
BC015695.2 | - | 6 | 1374 | 4842665 | ||
BC015779.1 | - | 6 | 1348 | 4856294 | ||
BC017450.1 | - | 5 | 1279 | |||
BC018774.1 | - | 6 | 1372 | 4869628 | ||
BC018861.2 | - | 3 | 812 | |||
BC023548.2 | - | 4 | 979 | |||
BC053572.1 | - | 6 | 1365 | 3865767 | ||
BC063495.1 | - | 5 | 1304 | |||
EU668331.1 | - | 3 | 884 | |||
M16247.1 | - | 3 | 884 | |||
X04098.1 | - | 6 | 1386 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
