siRNA Details
Gene Symbol: C14orf180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008404.2 | NP_001008404.1 | 5 | 1066 | ||
NM_001286399.1 | NP_001273328.1 | 5 | 1022 | |||
NM_001286400.1 | NP_001273329.1 | 5 | 1231 | |||
XM_005267638.3 | XP_005267695.1 | 5 | 1135 | |||
XM_011536764.2 | XP_011535066.1 | 5 | 1113 | |||
GenBank mRNA | AK095913.1 | - | 5 | 1031 | ||
AK297818.1 | - | 5 | 933 | |||
BX161457.1 | - | 5 | 1231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
