siRNA Details
Gene Symbol: RAB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193532.1 | NP_001180461.1 | 1 | 175 | ||
XM_017000226.1 | XP_016855715.1 | 1 | 190 | |||
GenBank mRNA | CX788289.1 | - | 1 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
