siRNA Details
Gene Symbol: SETD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007259.2 | NP_001007260.1 | 7 | 992 | ||
NM_001007261.2 | NP_001007262.1 | 8 | 1086 | |||
NM_001286752.1 | NP_001273681.1 | 8 | 1475 | |||
NM_017438.4 | NP_059134.1 | 7 | 1332 | |||
NR_040087.2 | 6 | 1243 | ||||
XM_011529636.1 | XP_011527938.1 | 7 | 1381 | |||
XM_011529637.1 | XP_011527939.1 | 7 | 1446 | |||
XM_011529638.2 | XP_011527940.1 | 7 | 1354 | |||
XM_011529639.1 | XP_011527941.1 | 7 | 1132 | |||
XM_011529640.2 | XP_011527942.1 | 7 | 1397 | |||
XM_011529642.1 | XP_011527944.1 | 8 | 1475 | |||
XM_011529643.1 | XP_011527945.1 | 8 | 1426 | |||
XM_011529644.1 | XP_011527946.1 | 8 | 987 | |||
XM_017028403.1 | XP_016883892.1 | 7 | 923 | |||
XM_017028404.1 | XP_016883893.1 | 8 | 1031 | |||
XM_017028405.1 | XP_016883894.1 | 7 | 1332 | |||
GenBank mRNA | AB004848.1 | - | 8 | 1039 | ||
AB004849.1 | - | 2 | 542 | |||
AB004852.1 | - | 6 | 863 | |||
AB004853.1 | - | 8 | 1035 | |||
AF391112.1 | - | 6 | 810 | |||
AK001660.1 | - | 7 | 899 | |||
AK095215.1 | - | 6 | 2199 | |||
AK300009.1 | - | 6 | 943 | |||
BC002898.2 | - | 7 | 931 | 3940849 | ||
BC036556.2 | - | 8 | 1149 | 5266627 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
