siRNA Details
Gene Symbol: HIST1H2AH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080596.2 | NP_542163.1 | 1 | 65 | ||
GenBank mRNA | AK312232.1 | - | 1 | 41 | ||
BC093849.1 | - | 1 | 65 | 7939694 | ||
BC093851.1 | - | 1 | 65 | 7939696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
