siRNA Details
Gene Symbol: OR4F15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001674.1 | NP_001001674.1 | 1 | 202 | ||
GenBank mRNA | BC137064.1 | - | 1 | 226 | 9021061 | |
BC137065.1 | - | 1 | 226 | 9021062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
