siRNA Details
Gene Symbol: OR4F15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001674.1 | NP_001001674.1 | 1 | 248 | ||
GenBank mRNA | BC137064.1 | - | 1 | 272 | 9021061 | |
BC137065.1 | - | 1 | 272 | 9021062 |
Gene Symbol: OR4F13P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046417.1 | 5 | 623 | |||
GenBank mRNA | AY792621.1 | - | 5 | 623 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
