siRNA Details
Gene Symbol: PIWIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008496.3 | NP_001008496.2 | 16 | 2387 | ||
NM_001255975.1 | NP_001242904.1 | 16 | 2360 | |||
NR_045648.1 | 17 | 2570 | ||||
NR_045649.1 | 17 | 2443 | ||||
GenBank mRNA | AB079368.1 | - | 16 | 2383 | ||
AK302409.1 | - | 16 | 2360 | |||
AK315806.1 | - | 16 | 2360 | |||
BC144693.1 | - | 17 | 2200 | |||
BC144696.1 | - | 17 | 2327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
