siRNA Details
Gene Symbol: PIWIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008496.3 | NP_001008496.2 | 18 | 2559 | ||
NM_001255975.1 | NP_001242904.1 | 18 | 2532 | |||
NR_045648.1 | 19 | 2742 | ||||
NR_045649.1 | 19 | 2615 | ||||
GenBank mRNA | AB079368.1 | - | 18 | 2555 | ||
AK302409.1 | - | 18 | 2532 | |||
AK315806.1 | - | 18 | 2532 | |||
BC144693.1 | - | 19 | 2372 | |||
BC144696.1 | - | 19 | 2499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
